Assay Details
| Gene Symbol: NAIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193947.2 | NP_001180876.1 | 653 | 112 | ||
| XM_015116588.2 | XP_014972074.1 | 761 | 112 | |||
| GenBank mRNA | JV635642.1 | - | 647 | 112 | ||
More Information
Set Membership: |
|

NAIF1
Genomic Map