Assay Details
| Gene Symbol: TMEM88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194489.2 | NP_001181418.1 | 234 | 67 | ||
| GenBank mRNA | JV044328.1 | - | 221 | 67 | ||
| JV635743.1 | - | 234 | 67 | |||
More Information
Set Membership: |
|

TMEM88
Genomic Map