Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098649.3 | XP_001098649.1 | 2185 | 96 | ||
| XM_028844523.1 | XP_028700356.1 | 2053 | 96 | |||
More Information
Set Membership: |
|

KBTBD12
Genomic Map