Assay Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837389.1 | XP_028693222.1 | 1159 | 130 | ||
| XM_028837390.1 | XP_028693223.1 | 970 | 130 | |||
| GenBank mRNA | JV043555.1 | - | 710 | 130 | ||
| JV635772.1 | - | 713 | 130 | |||
More Information
Set Membership: |
|

DCUN1D2
Genomic Map