Assay Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194584.2 | NP_001181513.2 | 382 | 92 | ||
| XM_015141285.2 | XP_014996771.2 | 291 | 92 | |||
| GenBank mRNA | JU323276.1 | - | 346 | 92 | ||
| JU473319.1 | - | 358 | 92 | |||
More Information
Set Membership: |
|

MRPL22
Genomic Map