Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082856.4 | XP_001082856.1 | 1435 | 63 | ||
| XM_015114701.2 | XP_014970187.1 | 1292 | 63 | |||
More Information
Set Membership: |
|

TRIM17
Genomic Map