Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092156.4 | XP_001092156.3 | 1121 | 104 | ||
| XM_015117895.2 | XP_014973381.1 | 559 | 104 | |||
| XM_028835240.1 | XP_028691073.1 | 704 | 104 | |||
| XM_028835241.1 | XP_028691074.1 | 1281 | 104 | |||
| GenBank mRNA | JU320828.1 | - | 996 | 104 | ||
| JU320829.1 | - | 996 | 104 | |||
| JU475821.1 | - | 841 | 104 | |||
| JV043814.1 | - | 790 | 104 | |||
More Information
Set Membership: |
|

ABHD17B
Genomic Map