Assay Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145783.2 | XP_015001269.2 | 2609 | 100 | ||
| GenBank mRNA | JU485005.1 | - | 74 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO43| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145783.2 | XP_015001269.2 | 2609 | 100 | ||
| GenBank mRNA | JU485005.1 | - | 74 | 100 | ||
Set Membership: |
|