Assay Details
Gene Symbol: MDH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106735.4 | XP_001106735.1 | 1307 | 122 | ||
XM_015110790.2 | XP_014966276.1 | 1306 | 122 | |||
XM_015110797.2 | XP_014966283.1 | 1180 | 122 | |||
XM_028831040.1 | XP_028686873.1 | 1306 | 122 | |||
XM_028831041.1 | XP_028686874.1 | 1306 | 122 | |||
XM_028831042.1 | XP_028686875.1 | 1306 | 122 | |||
XM_028831043.1 | XP_028686876.1 | 1306 | 122 | |||
XM_028831044.1 | XP_028686877.1 | 1009 | 122 | |||
XM_028831045.1 | XP_028686878.1 | 1306 | 122 | |||
XM_028831046.1 | XP_028686879.1 | 1306 | 122 | |||
XM_028831047.1 | XP_028686880.1 | 1198 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
