Assay Details
| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152464.2 | XP_015007950.2 | 2394 | 118 | ||
| XM_015152465.2 | XP_015007951.2 | 2116 | 118 | |||
| XM_015152466.2 | XP_015007952.2 | 1911 | 118 | |||
| XM_028829787.1 | XP_028685620.1 | 1772 | 118 | |||
| GenBank mRNA | JU349354.1 | - | 1410 | 118 | ||
| JU477461.1 | - | 1834 | 118 | |||
More Information
Set Membership: |
|

USP44
Genomic Map