Assay Details
| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040029.1 | NP_001035118.1 | 426 | 98 | ||
| GenBank mRNA | DQ341470.1 | - | 426 | 98 | ||
| JV651490.1 | - | 267 | 98 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EPCAM
Genomic Map