Assay Details
| Gene Symbol: MFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150286.2 | XP_015005772.1 | 2356 | 121 | ||
| GenBank mRNA | JU323965.1 | - | 738 | 121 | ||
| JU471508.1 | - | 674 | 121 | |||
More Information
Set Membership: |
|

MFNG
Genomic Map