Assay Details
| Gene Symbol: MAP3K10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123970.2 | XP_014979456.1 | 1787 | 65 | ||
| GenBank mRNA | JU324262.1 | - | 1468 | 65 | ||
| JV565122.1 | - | 1367 | 65 | |||
| JV684451.1 | - | 1471 | 65 | |||
| JV715323.1 | - | 1470 | 65 | |||
More Information
Set Membership: |
|

MAP3K10
Genomic Map