Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261734.1 | NP_001248663.1 | 670 | 155 | ||
| GenBank mRNA | JU324069.1 | - | 934 | 155 | ||
| JV045334.1 | - | 670 | 155 | |||
| JV625577.1 | - | 919 | 155 | |||
More Information
Set Membership: |
|

SFRP1
Genomic Map