Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130676.2 | XP_014986162.1 | 1667 | 80 | ||
| XM_015130677.2 | XP_014986163.1 | 771 | 80 | |||
| GenBank mRNA | JU328423.1 | - | 171 | 80 | ||
More Information
Set Membership: |
|

WNT5A
Genomic Map