Assay Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132684.2 | XP_014988170.2 | 269 | 93 | ||
| XM_015132702.2 | XP_014988188.2 | 143 | 93 | |||
| XM_015132712.2 | XP_014988198.2 | 175 | 93 | |||
| GenBank mRNA | JU325474.1 | - | 222 | 93 | ||
| JV565021.1 | - | 237 | 93 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015132702.2, XM_015132712.2 transcripts.
This assay is in the 5'UTR region of the XM_015132702.2, XM_015132712.2 transcripts.
Set Membership: |
|

FCN3
Genomic Map