Assay Details
| Gene Symbol: ARSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126895.2 | XP_014982381.2 | 577 | 104 | ||
| XM_028842198.1 | XP_028698031.1 | 578 | 104 | |||
| XM_028842199.1 | XP_028698032.1 | 608 | 104 | |||
| XM_028842200.1 | XP_028698033.1 | 602 | 104 | |||
More Information
Set Membership: |
|

ARSF
Genomic Map