Assay Details
| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121629.2 | XP_014977115.2 | 671 | 113 | ||
| GenBank mRNA | JU494645.1 | - | 239 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMILIN2| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121629.2 | XP_014977115.2 | 671 | 113 | ||
| GenBank mRNA | JU494645.1 | - | 239 | 113 | ||
Set Membership: |
|