Assay Details
| Gene Symbol: SLC7A6OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126469.2 | XP_014981955.1 | 714 | 92 | ||
| GenBank mRNA | JV726479.1 | - | 3959 | 92 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC7A6OS
Genomic Map