Assay Details
| Gene Symbol: SLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125424.2 | XP_014980910.2 | 5798 | 63 | ||
| XM_028840974.1 | XP_028696807.1 | 5795 | 63 | |||
| XM_028840975.1 | XP_028696808.1 | 4775 | 63 | |||
More Information
Set Membership: |
|

SLX4
Genomic Map