Assay Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099412.3 | XP_001099412.2 | 765 | 125 | ||
| XM_001099616.3 | XP_001099616.2 | 545 | 125 | |||
| GenBank mRNA | JV635535.1 | - | 619 | 125 | ||
| JV635536.1 | - | 601 | 125 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDGFD
Genomic Map