Assay Details
| Gene Symbol: CELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193955.3 | NP_001180884.2 | 826 | 90 | ||
| XM_015142598.2 | XP_014998084.2 | 590 | 90 | |||
| XM_015142599.2 | XP_014998085.2 | 589 | 90 | |||
| XM_015142600.2 | XP_014998086.2 | 588 | 90 | |||
| XM_015142601.2 | XP_014998087.2 | 588 | 90 | |||
| XM_015142602.2 | XP_014998088.2 | 590 | 90 | |||
| XM_015142603.2 | XP_014998089.2 | 593 | 90 | |||
| XM_015142606.2 | XP_014998092.1 | 464 | 90 | |||
| XM_028850240.1 | XP_028706073.1 | 957 | 90 | |||
| GenBank mRNA | JV589939.1 | - | 488 | 90 | ||
| JV624943.1 | - | 514 | 90 | |||
More Information
Set Membership: |
|

CELF6
Genomic Map