Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141247.2 | XP_014996733.1 | 474 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015141247.2 transcripts.
Set Membership: |
|

ZNF300
Genomic Map