Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101327.4 | XP_001101327.2 | 1164 | 113 | ||
| GenBank mRNA | JV706286.1 | - | 555 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED9| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101327.4 | XP_001101327.2 | 1164 | 113 | ||
| GenBank mRNA | JV706286.1 | - | 555 | 113 | ||
Set Membership: |
|