Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278397.1 | NP_001265326.1 | 1116 | 136 | ||
| XM_015115026.2 | XP_014970512.2 | 992 | 136 | |||
| XM_015115027.2 | XP_014970513.2 | 991 | 263 | |||
| XM_015115028.2 | XP_014970514.2 | 999 | 260 | |||
| XM_015115031.2 | XP_014970517.2 | 989 | 136 | |||
| XM_015115032.2 | XP_014970518.2 | 1010 | 136 | |||
| XM_015115033.2 | XP_014970519.2 | 1123 | 136 | |||
| XM_028832496.1 | XP_028688329.1 | 1415 | 260 | |||
| XM_028832498.1 | XP_028688331.1 | 846 | 136 | |||
More Information
Set Membership: |
|

ART5
Genomic Map