Assay Details
| Gene Symbol: CLIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087698.4 | XP_001087698.2 | 1995 | 139 | ||
| GenBank mRNA | JV584808.1 | - | 262 | 139 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLIC6| Gene Symbol: CLIC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087698.4 | XP_001087698.2 | 1995 | 139 | ||
| GenBank mRNA | JV584808.1 | - | 262 | 139 | ||
Set Membership: |
|