Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116196.2 | XP_014971682.1 | 1252 | 89 | ||
| XM_015116197.2 | XP_014971683.1 | 1269 | 89 | |||
| XM_028834193.1 | XP_028690026.1 | 1091 | 89 | |||
| GenBank mRNA | JV565820.1 | - | 1109 | 89 | ||
More Information
Set Membership: |
|

B3GAT1
Genomic Map