Assay Details
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266737.1 | NP_001253666.1 | 1350 | 80 | ||
XM_015128096.2 | XP_014983582.1 | 1557 | 80 | |||
XM_028841653.1 | XP_028697486.1 | 1458 | 80 | |||
XM_028841654.1 | XP_028697487.1 | 1419 | 80 | |||
GenBank mRNA | JU326640.1 | - | 557 | 80 | ||
JV565007.1 | - | 1350 | 80 | |||
JV721455.1 | - | 553 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
