Assay Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099038.4 | XP_001099038.2 | 596 | 136 | ||
| XM_028849814.1 | XP_028705647.1 | 629 | 136 | |||
More Information
Set Membership: |
|

PRELID2
Genomic Map