Assay Details
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260714.1 | NP_001247643.1 | 87 | 128 | ||
GenBank mRNA | JU323435.1 | - | 86 | 128 | ||
JU472067.1 | - | 72 | 128 | |||
JV048641.1 | - | 87 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260714.1 transcripts.
Set Membership: |
![]() ![]() |
