Assay Details
| Gene Symbol: VIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265773.1 | NP_001252702.1 | 964 | 67 | ||
| XM_015131181.2 | XP_014986667.1 | 951 | 67 | |||
| XM_028843435.1 | XP_028699268.1 | 955 | 67 | |||
| GenBank mRNA | JV556807.1 | - | 862 | 67 | ||
| JV565010.1 | - | 964 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VIPR1
Genomic Map