Assay Details
| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087096.4 | XP_001087096.2 | 1389 | 88 | ||
| XM_015131341.2 | XP_014986827.2 | 714 | 88 | |||
| XM_015131343.2 | XP_014986829.2 | 714 | 88 | |||
| XM_015131344.2 | XP_014986830.2 | 714 | 88 | |||
| XM_015131345.2 | XP_014986831.2 | 1337 | 88 | |||
| XM_015131346.2 | XP_014986832.2 | 1416 | 88 | |||
| XM_015131348.2 | XP_014986834.2 | 614 | 88 | |||
| XM_015131349.2 | XP_014986835.2 | 598 | 88 | |||
| GenBank mRNA | JV727192.1 | - | 288 | 88 | ||
More Information
Set Membership: |
|

SLC22A14
Genomic Map