Assay Details
| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001081975.4 | XP_001081975.2 | 215 | 114 | ||
| XM_001082215.4 | XP_001082215.1 | 325 | 114 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MB| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001081975.4 | XP_001081975.2 | 215 | 114 | ||
| XM_001082215.4 | XP_001082215.1 | 325 | 114 | |||
Set Membership: |
|