Assay Details
| Gene Symbol: TAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194446.2 | NP_001181375.1 | 281 | 125 | ||
| XM_015117088.2 | XP_014972574.1 | 325 | 125 | |||
| XM_028834363.1 | XP_028690196.1 | 355 | 125 | |||
| GenBank mRNA | JU336111.1 | - | 261 | 125 | ||
More Information
Set Membership: |
|

TAL2
Genomic Map