Assay Details
| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100890.4 | XP_001100890.4 | 487 | 85 | ||
| XM_028828345.1 | XP_028684178.1 | 511 | 85 | |||
| XM_028828346.1 | XP_028684179.1 | 508 | 85 | |||
| XM_028828347.1 | XP_028684180.1 | 508 | 85 | |||
| GenBank mRNA | JU327550.1 | - | 238 | 85 | ||
| JV531543.1 | - | 310 | 85 | |||
More Information
Set Membership: |
|

MMP24
Genomic Map