Assay Details
| Gene Symbol: LYPLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266699.1 | NP_001253628.1 | 409 | 68 | ||
| XM_028846084.1 | XP_028701917.1 | 1384 | 68 | |||
| GenBank mRNA | JU320872.1 | - | 398 | 68 | ||
| JU471506.1 | - | 409 | 68 | |||
| JV638622.1 | - | 411 | 68 | |||
More Information
Set Membership: |
|

LYPLA2
Genomic Map