Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194081.2 | NP_001181010.2 | 230 | 71 | ||
| GenBank mRNA | JU322752.1 | - | 136 | 71 | ||
| JU474979.1 | - | 205 | 71 | |||
| JV710981.1 | - | 146 | 71 | |||
More Information
Set Membership: |
|

HEBP2
Genomic Map