Assay Details
| Gene Symbol: SIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267025.1 | NP_001253954.1 | 391 | 94 | ||
| GenBank mRNA | JU475196.1 | - | 391 | 94 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SIT1
Genomic Map