Assay Details
Gene Symbol: GARRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193886.2 | NP_001180815.1 | 3680 | 135 | ||
XM_015123517.2 | XP_014979003.1 | 5236 | 135 | |||
XM_015123518.2 | XP_014979004.1 | 3867 | 135 | |||
XM_028838424.1 | XP_028694257.1 | 3827 | 135 | |||
XM_028838426.1 | XP_028694259.1 | 3249 | 135 | |||
XM_028838427.1 | XP_028694260.1 | 3306 | 135 | |||
GenBank mRNA | JU321568.1 | - | 3669 | 135 | ||
JU470676.1 | - | 3666 | 135 | |||
JV045078.1 | - | 3560 | 135 | |||
JV636941.1 | - | 3662 | 135 |

More Information
Set Membership: |
![]() ![]() |
