Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266395.1 | NP_001253324.1 | 1240 | 79 | ||
| XM_015119236.2 | XP_014974722.1 | 1204 | 79 | |||
| XM_028835492.1 | XP_028691325.1 | 1201 | 79 | |||
| XM_028835493.1 | XP_028691326.1 | 1216 | 79 | |||
| GenBank mRNA | JU340485.1 | - | 1212 | 79 | ||
| JU471697.1 | - | 1240 | 79 | |||
More Information
Set Membership: |
|

ZNF652
Genomic Map