Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116763.4 | XP_001116763.4 | 1665 | 75 | ||
| XM_028847620.1 | XP_028703453.1 | 1213 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETV7| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116763.4 | XP_001116763.4 | 1665 | 75 | ||
| XM_028847620.1 | XP_028703453.1 | 1213 | 75 | |||
Set Membership: |
|