Assay Details
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097842.4 | XP_001097842.1 | 699 | 65 | ||
| XM_015139709.2 | XP_014995195.1 | 699 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRX4| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097842.4 | XP_001097842.1 | 699 | 65 | ||
| XM_015139709.2 | XP_014995195.1 | 699 | 65 | |||
Set Membership: |
|