Assay Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116963.4 | XP_001116963.2 | 1127 | 78 | ||
| XM_015120195.2 | XP_014975681.1 | 1127 | 78 | |||
More Information
Set Membership: |
|

FAM20A
Genomic Map