Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089622.4 | XP_001089622.2 | 406 | 105 | ||
| XM_015120280.2 | XP_014975766.2 | 406 | 105 | |||
| GenBank mRNA | JU335135.1 | - | 405 | 105 | ||
| JV043504.1 | - | 336 | 105 | |||
More Information
Set Membership: |
|

TMEM104
Genomic Map