Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080135.1 | NP_001073604.1 | 166 | 105 | ||
| GenBank mRNA | EF121759.1 | - | 166 | 105 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ODAM
Genomic Map