Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194325.1 | NP_001181254.1 | 230 | 85 | ||
| GenBank mRNA | JU322549.1 | - | 220 | 85 | ||
| JV637314.1 | - | 228 | 85 | |||
More Information
Set Membership: |
|

TMEM38B
Genomic Map