Assay Details
| Gene Symbol: SMG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194385.1 | NP_001181314.1 | 1801 | 127 | ||
| GenBank mRNA | JU335254.1 | - | 1759 | 127 | ||
| JU489100.1 | - | 1747 | 127 | |||
| JV586108.1 | - | 870 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMG8
Genomic Map