Assay Details
| Gene Symbol: SMG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194385.1 | NP_001181314.1 | 1950 | 129 | ||
| GenBank mRNA | JU335254.1 | - | 1908 | 129 | ||
| JU489100.1 | - | 1896 | 129 | |||
| JV586108.1 | - | 1019 | 129 | |||
More Information
Set Membership: |
|

SMG8
Genomic Map