Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095451.4 | XP_001095451.3 | 491 | 169 | ||
| GenBank mRNA | JV622887.1 | - | 324 | 168 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAAT1| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095451.4 | XP_001095451.3 | 491 | 169 | ||
| GenBank mRNA | JV622887.1 | - | 324 | 168 | ||
Set Membership: |
|