Assay Details
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144415.2 | XP_014999901.2 | 2601 | 98 | ||
GenBank mRNA | JU335562.1 | - | 2353 | 98 | ||
JV513706.1 | - | 2311 | 98 | |||
JV636282.1 | - | 5216 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
